ContactUsLinkComponent
Mouse C5a, His Tag (C5A-M51H4) is expressed from E.coli cells. It contains AA Asn 679 - Arg 755 (Accession P06684). 1 * 100 µG
Catalog # ACRBC5A-M51H4100UG
Supplier:  ACRO Biosystems
Mouse C5a, His Tag (C5A-M51H4) is expressed from E.coli cells. It contains AA Asn 679 - Arg 755 (Accession P06684). 1 * 100 µG
Catalog # ACRBC5A-M51H4100UG
Supplier:  ACRO Biosystems
Frequently Bought Together