ContactUsLinkComponent
Human Neuropilin-1, His Tag (NR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Lys 644 (Accession AAH07 533). 1 * 50 µG
Catalog # ACRBNR1-H5228-50UG
Supplier:  ACRO Biosystems
Human Neuropilin-1, His Tag (NR1-H5228) is expressed from human 293 cells (HEK293). It contains AA Phe 22 - Lys 644 (Accession AAH07 533). 1 * 50 µG
Catalog # ACRBNR1-H5228-50UG
Supplier:  ACRO Biosystems
Frequently Bought Together