ContactUsLinkComponent
Biotinylated Human CTLA-4, His,Avitag (CT4-H82E1) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Phe 162 (Acces sion Q6GR94-1). 1 * 25 µG
Catalog # ACRBCT4-H82E1-25UG
Supplier:  ACRO Biosystems
Biotinylated Human CTLA-4, His,Avitag (CT4-H82E1) is expressed from human 293 cells (HEK293). It contains AA Ala 37 - Phe 162 (Acces sion Q6GR94-1). 1 * 25 µG
Catalog # ACRBCT4-H82E1-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together