Order Entry
Switzerland
Orders LinkContactUsLinkComponent
Rabbit recombinant Complement C5 (from HEK293 cells)

Rabbit recombinant Complement C5 (from HEK293 cells)

1 mg
Catalog # ACRBCO5-R52H5-1MG
Marketsource
Supplier:   ACRO Biosystems
Rabbit recombinant Complement C5 (from HEK293 cells)
Catalog # ACRBCO5-R52H5-1MG
Supplier:  ACRO Biosystems
Marketsource
CAS Number:  
Rabbit recombinant Complement C5 (from HEK293 cells)

1 mg

CAS Number:  
Price
2 615.00 SFr.
Log in for availability
1 * 1 mg
Product Details & Documents

Rabbit Complement C5 protein, His Tag.

Specifications

Specifications

  • Pk:
    1 mg
  • Protein/peptide name:
    Complement C5
  • Protein synonyms:
    CPAMD4|Complement C5|C5
  • Purity:
    >90% as determined by SDS-PAGE.
  • Endotoxin level:
    Low
  • Source:
    HEK293 cells
  • Molecular weight:
    73,8 kDa (β chain) and 111,9 kDa (α chain)
  • Shipping temperature:
    RT
  • Cat. No.:
    ACRBCO5-R52H5-1MG
  • Protein/peptide type:
    Recombinant
  • Endotoxin content:
    Less than 1,0 EU per μg by the LAL method.
  • Storage conditions:
    –20 °C
  • Formulation:
    Lyophilised from 0,22 μm filtered solution in PBS, pH 7,4. Normally trehalose is added as protectant before Lyophilisation.
  • Conjugation:
    Unconjugated
  • Tag sequence:
    His Tag
  • Species:
    Rabbit

Specifications

Empfehlungen

Empfehlungen werden nur dann basierend auf Ihren Einkaufspräferenzen personalisiert, wenn Sie auf der Seite "Persönliche Daten" die Option „Mein Einkaufserlebnis verbessern“ aktiviert haben.

Andernfalls erhalten Sie allgemeine Empfehlungen..