ContactUsLinkComponent
Recombinat Protein,Mouse BTLA Protein, Fc Tag,Mouse BTLA, Fc Tag (BTA-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Gly 176 (Accession Q32MV9).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a 1 * 1 mg
Catalog # ACRBBTA-M5253-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Mouse BTLA Protein, Fc Tag,Mouse BTLA, Fc Tag (BTA-M5253) is expressed from human 293 cells (HEK293). It contains AA Glu 30 - Gly 176 (Accession Q32MV9).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has a 1 * 1 mg
Catalog # ACRBBTA-M5253-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together