ContactUsLinkComponent
Recombinat Protein,Human VSIG4 Protein, His Tag,Human VSIG4, His Tag (VS4-H5226) is expressed from human 293 cells (HEK293). It cont ains AA Arg 20 - Pro 283 (Accession AAH10525).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 100 µG
Catalog # ACRBVS4-H5226100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human VSIG4 Protein, His Tag,Human VSIG4, His Tag (VS4-H5226) is expressed from human 293 cells (HEK293). It cont ains AA Arg 20 - Pro 283 (Accession AAH10525).This protein carries a polyhistidine tag at the C-terminus. The protein 1 * 100 µG
Catalog # ACRBVS4-H5226100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together