ContactUsLinkComponent
Biotinylated Human IL-15 R alpha, Fc,Avitag (ILA-H82F4) is expressed from human 293 cells (HEK293). It contains AA Ile 31 - Thr 205 (Accession Q13261-1). 1 * 25 µG
Catalog # ACRBILA-H82F4-25UG
Supplier:  ACRO Biosystems
Biotinylated Human IL-15 R alpha, Fc,Avitag (ILA-H82F4) is expressed from human 293 cells (HEK293). It contains AA Ile 31 - Thr 205 (Accession Q13261-1). 1 * 25 µG
Catalog # ACRBILA-H82F4-25UG
Supplier:  ACRO Biosystems
Frequently Bought Together