ContactUsLinkComponent
Biotinylated Cynomolgus CD32b, His,Avitag (SPR BLI verified) (CDB-C82E4) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 224 (Accession Q8SPW3-1). 1 * 200 µG
Catalog # ACRBCDB-C82E4-200
Supplier:  ACRO Biosystems
Biotinylated Cynomolgus CD32b, His,Avitag (SPR BLI verified) (CDB-C82E4) is expressed from human 293 cells (HEK293). It contains AA Ala 46 - Pro 224 (Accession Q8SPW3-1). 1 * 200 µG
Catalog # ACRBCDB-C82E4-200
Supplier:  ACRO Biosystems
Frequently Bought Together