Biotinylated Cynomolgus CD64, His,Avitag (BLI verified) (FCA-C82E8) is expressed from human 293 cells (HEK293). It contains AA Val 1 1 - Pro 288 (Accession NP001270969.1). 1 * 200 µG
Katalognr. ACRBFCA-C82E8-200
Anbieter: ACRO Biosystems
Frequently Bought Together