ContactUsLinkComponent
Recombinat Protein,Human P-Selectin CD62P Protein, Fc Tag,Human P-Selectin, Fc Tag (SEP-H5255) is expressed from human 293 cells (HE K293). It contains AA Trp 42 - Ala 771 (Accession AAN06828).This protein carries a human IgG1 Fc tag at the C-terminu 1 * 100 µG
Catalog # ACRBSEP-H5255100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human P-Selectin CD62P Protein, Fc Tag,Human P-Selectin, Fc Tag (SEP-H5255) is expressed from human 293 cells (HE K293). It contains AA Trp 42 - Ala 771 (Accession AAN06828).This protein carries a human IgG1 Fc tag at the C-terminu 1 * 100 µG
Catalog # ACRBSEP-H5255100UG
Supplier:  ACRO Biosystems
Frequently Bought Together