ContactUsLinkComponent
Unconjugated Human PDGF-BB, His,Avitag (PDB-H5127) is expressed from E.coli cells. It contains AA Ser 82 - Thr 190 (Accession P01127 -1). 1 * 200 µG
Catalog # ACRBPDB-H5127200UG
Supplier:  ACRO Biosystems
Unconjugated Human PDGF-BB, His,Avitag (PDB-H5127) is expressed from E.coli cells. It contains AA Ser 82 - Thr 190 (Accession P01127 -1). 1 * 200 µG
Catalog # ACRBPDB-H5127200UG
Supplier:  ACRO Biosystems
Frequently Bought Together