ContactUsLinkComponent
Recombinat Protein,Human P-Selectin CD62P Protein, His Tag,Human P-Selectin, His Tag (SEP-H52E4) is expressed from human 293 cells ( HEK293). It contains AA Trp 42 - Ala 771 (Accession AAN06828).This protein carries a polyhistidine tag at the C-termi 1 * 100 µG
Catalog # ACRBSEP-H52E4100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Human P-Selectin CD62P Protein, His Tag,Human P-Selectin, His Tag (SEP-H52E4) is expressed from human 293 cells ( HEK293). It contains AA Trp 42 - Ala 771 (Accession AAN06828).This protein carries a polyhistidine tag at the C-termi 1 * 100 µG
Catalog # ACRBSEP-H52E4100UG
Supplier:  ACRO Biosystems
Frequently Bought Together