ContactUsLinkComponent
Human PVRIG, Fc Tag (PVG-H5257) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession Q6DKI7-1). 1 * 100 µG
Catalog # ACRBPVG-H5257100UG
Supplier:  ACRO Biosystems
Human PVRIG, Fc Tag (PVG-H5257) is expressed from human 293 cells (HEK293). It contains AA Thr 41 - Asp 171 (Accession Q6DKI7-1). 1 * 100 µG
Catalog # ACRBPVG-H5257100UG
Supplier:  ACRO Biosystems
Frequently Bought Together