ContactUsLinkComponent
Human BAFFR, Llama IgG2b Fc Tag, low endotoxin (BAR-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession Q96RJ3-1). 1 * 1 mg
Catalog # ACRBBAR-H5258-1MG
Supplier:  ACRO Biosystems
Human BAFFR, Llama IgG2b Fc Tag, low endotoxin (BAR-H5258) is expressed from human 293 cells (HEK293). It contains AA Ser 7 - Ala 71 (Accession Q96RJ3-1). 1 * 1 mg
Catalog # ACRBBAR-H5258-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together