Recombinat Protein,Human IL-17C Protein, His Tag,Human IL-17C, His Tag (ILC-H52H7) is expressed from human 293 cells (HEK293). It co ntains AA His 19 - Val 197 (Accession Q9P0M4-1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 1 mg
Katalognr. ACRBILC-H52H7-1MG
Supplier: ACRO Biosystems
Frequently Bought Together