ContactUsLinkComponent
Human FOLR2, His Tag (FO2-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 17 - His 228 (Accession P14207-1). 1 * 1 mg
Catalog # ACRBFO2-H5223-1MG
Supplier:  ACRO Biosystems
Human FOLR2, His Tag (FO2-H5223) is expressed from human 293 cells (HEK293). It contains AA Thr 17 - His 228 (Accession P14207-1). 1 * 1 mg
Catalog # ACRBFO2-H5223-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together