ContactUsLinkComponent
Biotinylated Human IL-13, His,Avitag (IL3-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Asn 132 (Access ion AAK53823.1). 1 * 200 µG
Catalog # ACRBIL3-H82E5200UG
Supplier:  ACRO Biosystems
Biotinylated Human IL-13, His,Avitag (IL3-H82E5) is expressed from human 293 cells (HEK293). It contains AA Gly 21 - Asn 132 (Access ion AAK53823.1). 1 * 200 µG
Catalog # ACRBIL3-H82E5200UG
Supplier:  ACRO Biosystems
Frequently Bought Together