ContactUsLinkComponent
Recombinat Protein,Rhesus macaque Glypican 2 GPC2 Protein, His Tag,Rhesus macaque Glypican 2, His Tag (GP2-R52H9) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 552 (Accession XP001103286).This protein carries a polyhistidine 1 * 100 µG
Katalognr. ACRBGP2-R52H9100UG
Supplier:  ACRO Biosystems
Recombinat Protein,Rhesus macaque Glypican 2 GPC2 Protein, His Tag,Rhesus macaque Glypican 2, His Tag (GP2-R52H9) is expressed from human 293 cells (HEK293). It contains AA Ser 24 - Ser 552 (Accession XP001103286).This protein carries a polyhistidine 1 * 100 µG
Katalognr. ACRBGP2-R52H9100UG
Supplier:  ACRO Biosystems
Frequently Bought Together