Recombinat Protein,Human CD206 MMR Protein, His Tag,Human CD206, His Tag (CD6-H52H9) is expressed from human 293 cells (HEK293). It contains AA Leu 19 - Ala 1389 (Accession P22897-1).This protein carries a polyhistidine tag at the C-terminus. The pro 1 * 1 mg
Catalog # ACRBCD6-H52H9-1MG
Supplier: ACRO Biosystems
Frequently Bought Together