ContactUsLinkComponent
Recombinat Protein,Human CD14 Protein, Fc Tag,Human CD14, Fc Tag (CD4-H5252) is expressed from human 293 cells (HEK293). It contains AA Thr 20 - Cys 352 (Accession P08571-1).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has 1 * 1 mg
Catalog # ACRBCD4-H5252-1MG
Supplier:  ACRO Biosystems
Recombinat Protein,Human CD14 Protein, Fc Tag,Human CD14, Fc Tag (CD4-H5252) is expressed from human 293 cells (HEK293). It contains AA Thr 20 - Cys 352 (Accession P08571-1).This protein carries a human IgG1 Fc tag at the C-terminus. The protein has 1 * 1 mg
Catalog # ACRBCD4-H5252-1MG
Supplier:  ACRO Biosystems
Frequently Bought Together