ContactUsLinkComponent
Recombinat Protein,Mouse RENIN Protein, His Tag,Mouse RENIN, His Tag (REN-M5222) is expressed from human 293 cells (HEK293). It cont ains AA Leu 22 - Arg 402 (Accession NP112469.1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 100 µG
Catalog # ACRBREN-M5222100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Mouse RENIN Protein, His Tag,Mouse RENIN, His Tag (REN-M5222) is expressed from human 293 cells (HEK293). It cont ains AA Leu 22 - Arg 402 (Accession NP112469.1).This protein carries a polyhistidine tag at the C-terminus. The prote 1 * 100 µG
Catalog # ACRBREN-M5222100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together