ContactUsLinkComponent
Recombinat Protein,Biotinylated Human NKp46 NCR1 CD335 Protein, Fc,Avitag™,Biotinylated Human NKp46, Fc,Avitag (NC1-H82F9) is expres sed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession AAH64806).This protein carries a human 1 * 25 µG
Catalog # ACRBNC1-H82F9-25UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Biotinylated Human NKp46 NCR1 CD335 Protein, Fc,Avitag™,Biotinylated Human NKp46, Fc,Avitag (NC1-H82F9) is expres sed from human 293 cells (HEK293). It contains AA Gln 22 - Asn 254 (Accession AAH64806).This protein carries a human 1 * 25 µG
Catalog # ACRBNC1-H82F9-25UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together