ContactUsLinkComponent
Recombinat Protein,Human Noggin Protein, Fc Tag,Human Noggin, Fc Tag (NON-H5257) is expressed from human 293 cells (HEK293). It cont ains AA Gln 28 - Cys 232 (Accession NP005441).This protein carries a human IgG1 Fc tag at the C-terminus. The protein 1 * 100 µG
Catalog # ACRBNON-H5257100UG
Supplier:  ACROBIOSYSTEMS
Recombinat Protein,Human Noggin Protein, Fc Tag,Human Noggin, Fc Tag (NON-H5257) is expressed from human 293 cells (HEK293). It cont ains AA Gln 28 - Cys 232 (Accession NP005441).This protein carries a human IgG1 Fc tag at the C-terminus. The protein 1 * 100 µG
Catalog # ACRBNON-H5257100UG
Supplier:  ACROBIOSYSTEMS
Frequently Bought Together