ContactUsLinkComponent
Human IL-2 R beta, His Tag (SPR verified) (CD2-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (A ccession NP000869.1). 1 * 500 µG
Catalog # ACRBCD2-H5221500UG
Supplier:  ACRO Biosystems
Human IL-2 R beta, His Tag (SPR verified) (CD2-H5221) is expressed from human 293 cells (HEK293). It contains AA Ala 27 - Asp 239 (A ccession NP000869.1). 1 * 500 µG
Catalog # ACRBCD2-H5221500UG
Supplier:  ACRO Biosystems
Frequently Bought Together