ContactUsLinkComponent
Human PD-1, His Tag (HPLC verified) (PD1-H5221) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accessi on NP005009.2). 1 * 100 µG
  ACRBPD1-H5221100UG
Human PD-1, His Tag (HPLC verified) (PD1-H5221) is expressed from human 293 cells (HEK293). It contains AA Leu 25 - Gln 167 (Accessi on NP005009.2). 1 * 100 µG
  ACRBPD1-H5221100UG
Frequently Bought Together