ContactUsLinkComponent
The handheld probe (Ø15 x 400 mm) is especially suitable for mobile measurements of high temperatures. It measures humidity and temperature and calculates the dew/frost point. Applications Climatic and temperature chambers, dryers, air ducts Features 1 * 1 items
Catalog # ROTRHC2-HK40
The handheld probe (Ø15 x 400 mm) is especially suitable for mobile measurements of high temperatures. It measures humidity and temperature and calculates the dew/frost point. Applications Climatic and temperature chambers, dryers, air ducts Features 1 * 1 items
Catalog # ROTRHC2-HK40
Frequently Bought Together