ContactUsLinkComponent
3151210s HWS flat flange reactor, model ADAV, DN 120s volume 1.000 ml, outletvalve T 10 mm, heating mantle in- / outlet flange DN 15p +++flange acc. DIN 12.214+++ Customs No. 70172000 1 * 1 items
Catalog # HWSA3151210S
Supplier:  HWS LABORTECHNIK
3151210s HWS flat flange reactor, model ADAV, DN 120s volume 1.000 ml, outletvalve T 10 mm, heating mantle in- / outlet flange DN 15p +++flange acc. DIN 12.214+++ Customs No. 70172000 1 * 1 items
Catalog # HWSA3151210S
Supplier:  HWS LABORTECHNIK
Frequently Bought Together