ContactUsLinkComponent
SARSTEDT SCREW TUBE, 50 ML, 114 X 28 MM, POINTED BOTTOM, PP, WITH PRINTED LETTERING FIELD AND SCALING, WITH ASSEMBLED CLOSURE, 25 ST K./BEUTE
Catalog # MSPP62.548.004
SARSTEDT SCREW TUBE, 50 ML, 114 X 28 MM, POINTED BOTTOM, PP, WITH PRINTED LETTERING FIELD AND SCALING, WITH ASSEMBLED CLOSURE, 25 ST K./BEUTE
Catalog # MSPP62.548.004
Frequently Bought Together