ContactUsLinkComponent
Melting point meter MPM-H3, for capillaries 80x1.4x1.0 mm with one end closed, accuracy of measurement +/- 0.4C, reproducibility +/- 0.3C,, safety class IP 20, lens 20 diopter, with USB interface, for visual measuring method, temperature rang 1 * 1 items
Catalog # MARI7232233
Supplier:  MARIENFELD
Melting point meter MPM-H3, for capillaries 80x1.4x1.0 mm with one end closed, accuracy of measurement +/- 0.4C, reproducibility +/- 0.3C,, safety class IP 20, lens 20 diopter, with USB interface, for visual measuring method, temperature rang 1 * 1 items
Catalog # MARI7232233
Supplier:  MARIENFELD
Frequently Bought Together