ContactUsLinkComponent
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, both SMMMS 50g/m², one blue, one green, 90x90cm, DIN EN ISO 11607 1 * 50 items
Référence VEPA3FDOP383212
Fournisseur:  VP Medical Packaging
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, both SMMMS 50g/m², one blue, one green, 90x90cm, DIN EN ISO 11607 1 * 50 items
Référence VEPA3FDOP383212
Fournisseur:  VP Medical Packaging
Frequently Bought Together