0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, both SMMMS 50g/m², one blue, one green, 60x60cm, DIN EN ISO 11607 1 * 125 items
Catalog # VEPA3FDOP383206
Supplier:
VP Medical Packaging
Expand image
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, both SMMMS 50g/m², one blue, one green, 60x60cm, DIN EN ISO 11607 1 * 125 items
Catalog # VEPA3FDOP383206
Supplier:
VP Medical Packaging
Frequently Bought Together
Nitrile gloves, TouchNTuff® 92-600 / 92-605
Supplier: Ansell
Nitrile gloves, Kimtech™ Purple Nitrile™ Xtra™
Supplier: KIMBERLY CLARK
VWR®, Nitrile Gloves
Supplier: VWR Collection
VWR®, Laboratory Markers
Supplier: VWR Collection
Lead-free steam indicator tapes, Comply™
Supplier: 3M MEDICAL
VWR® Nitrile Light Long, Nitrile Gloves
Supplier: VWR Collection
Nitrile gloves, Kimtech™ Purple Nitrile™
Supplier: KIMBERLY CLARK
VWR® NITRILE LIGHT, Nitrile Gloves
Supplier: VWR Collection
VWR® Labels, Hazardous Substance, GHS Labels
Supplier: VWR Collection
VWR®, Latex Gloves
Supplier: VWR Collection