ContactUsLinkComponent
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, both SMMMS 43g/m², one blue, one green, 100x100cm, DIN EN ISO 116 07 1 * 50 items
Catalog # VEPA3FDOP381214
Stericlin sterilisation double wrap, 2 sheets sealed at the edges, both SMMMS 43g/m², one blue, one green, 100x100cm, DIN EN ISO 116 07 1 * 50 items
Catalog # VEPA3FDOP381214
Frequently Bought Together