ContactUsLinkComponent
Bag waste disposal, 6l , biohazard , PP ,red ,with indicator field ,300 x 500 x 0.050 mm 1 * 500 items
Catalog # RATI7003200
Supplier:  RATIOLAB
Bag waste disposal, 6l , biohazard , PP ,red ,with indicator field ,300 x 500 x 0.050 mm 1 * 500 items
Catalog # RATI7003200
Supplier:  RATIOLAB
Frequently Bought Together