ContactUsLinkComponent
Cryo-Boxes, PP, grid 8 x 8, Blue, 133 x 133 x 50/75 mm, combi-lid, numerically coded 1 * 5 items
Catalog # RATI5320214
Supplier:  RATIOLAB
Cryo-Boxes, PP, grid 8 x 8, Blue, 133 x 133 x 50/75 mm, combi-lid, numerically coded 1 * 5 items
Catalog # RATI5320214
Supplier:  RATIOLAB
Frequently Bought Together