SFr. 0.00
Order Entry
/
Products
Services
Digital Solutions
Applications
Support
About
Switzerland
ContactUsLinkComponent
SWAB-TUBE 16/100 PP W/APPLICATOR PP STER 1 * 1.000 items
BOET09.313.4104
:
BOETTGER PAUL
SWAB-TUBE 16/100 PP W/APPLICATOR PP STER 1 * 1.000 items
BOET09.313.4104
:
BOETTGER PAUL
Frequently Bought Together
Buffer solutions, Certipur®, colour coded, Supelco®
: Merck
Sterican® Single-Use Needles for Special Applications
: B. Braun
Syringes, 3-piece
: TERUMO
VWR®, Microscope Slides
: VWR Collection
BD Plastipak™ Syringes
: BD Medical
Single use syringes, 2-piece, Injekt® Solo
: B. Braun
Disposable syringes, Omnifix®
: B. Braun
CLASSIQSwabs™ Sterile Dry Fiber Wrapped Swabs
: Copan
BD Discardit™ II Syringes
: BD Medical
Fine dosage syringes, Injekt®-F Solo
: B. Braun
Single-use syringes, 3-piece, HENKE-JECT®
: HENKE SASS WOLF
Single-use syringes, 2-piece, HENKE-JECT®
: HENKE SASS WOLF