ContactUsLinkComponent
After 1 year lease, option to buy full package consisting of ChromSwordAuto® 5 + AutoRobust Windows? Software for automatic HPLC method development, improvement and robustness tests Contents: ChromSword? Auto 5 Developer, ChromSword? Auto 5 Scout, Ch 1 * 1 items
Catalog # HPLC908-0111
After 1 year lease, option to buy full package consisting of ChromSwordAuto® 5 + AutoRobust Windows? Software for automatic HPLC method development, improvement and robustness tests Contents: ChromSword? Auto 5 Developer, ChromSword? Auto 5 Scout, Ch 1 * 1 items
Catalog # HPLC908-0111
Frequently Bought Together