Parteck SI 200 (Sorbitol) EMPROVE ESSENTIAL Ph Eur,BP,JP,NF,E 420 1 * 2,5 kg
Catalog # 1.15079.2500
Supplier: MERCK PRODUCTION CHEMICALS
Parteck SI 200 (Sorbitol) EMPROVE ESSENTIAL Ph Eur,BP,JP,NF,E 420 1 * 2,5 kg
Catalog # 1.15079.2500
Supplier: MERCK PRODUCTION CHEMICALS
Frequently Bought Together
Acetone ≥99.5%, GPR RECTAPUR®
Supplier: VWR Chemicals
Acetone ≥99%, TECHNICAL
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
n-Hexane ≥95%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Ethanol absolute ≥99.5%, EMPLURA®, Supelco®
Supplier: Merck
Methanol ≥99.8%, HiPerSolv CHROMANORM® Reag. Ph. Eur., gradient grade for HPLC
Supplier: VWR Chemicals
Acetonitrile ≥99.9%, HiPerSolv CHROMANORM®, gradient grade for HPLC
Supplier: VWR Chemicals
Centrifugal filter units, Amicon® Ultra-15
Supplier: Merck
Water, HiPerSolv CHROMANORM® for LC-MS, suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
Ethanol 96% vol Ph. Eur.
Supplier: VWR Chemicals
Ethanol absolute ≥99.8%, AnalaR NORMAPUR® Reag. Ph. Eur., ACS analytical reagent
Supplier: VWR Chemicals
Acetone ≥99.8%, HiPerSolv CHROMANORM® for HPLC
Supplier: VWR Chemicals
XYLITOL NF 1 * 2,5 kg
Supplier: Spectrum Chemical