Orders LinkContactUsLinkComponent
Safety spectacles, Eiger™
Safety spectacles, Eiger™
Eiger™ safety spectacles, Black/green, Smoke
Catalog # JSPA1EIGGREEN23SKN
Supplier:   JSP
Safety spectacles, Eiger™
Catalog # JSPA1EIGGREEN23SKN
Supplier:  JSP
Safety spectacles, Eiger™
Safety spectacles, Eiger™
Eiger™ safety spectacles, Black/green, Smoke
Price
SFr. 9.30
Log in for availability
1 * 1 ST
Product Details & Documents
Sporty design with optimal curved lens and large field of vision.
  • Moulded browguard
  • Excellence coating
  • Optimal lateral coverage
Active style and sporty design. Unique Flexofit technology engineered into temple cushioning to enhance wearer comfort, and to ensure a non-slip, low pressure fit.
Excellence coating on the optimal curved lens, large field of vision, moulded browguard. Additional gasket and strap accessory pack available to deliver a perfect dust barrier.
Certifications: Meets EN 166:2001.
Specifications
  • Colour:
    Black/green
  • Description:
    Eiger™ safety spectacles
  • Seq:
    02
  • Lenses:
    Smoke