ContactUsLinkComponent
CHROMABOND QuEChERS Mix III, Clean-up, EN 15662, tubes 15 mL,1,05 g,BIG 1 * 100 items
Catalog # MANA730972.100
Supplier:  MACHEREY-NAGEL
CHROMABOND QuEChERS Mix III, Clean-up, EN 15662, tubes 15 mL,1,05 g,BIG 1 * 100 items
Catalog # MANA730972.100
Supplier:  MACHEREY-NAGEL
Frequently Bought Together