ContactUsLinkComponent
0.2 um PVDF with GF Prefilter, 96-well plate, 1.0 m, PVDF with GF Prefilter, 96-well plate, 1.0 ml, Omni Screen Filter Plates, 96-we ll plate 1 * 10 ST
Catalog # MSPPOMNPF02-GF
0.2 um PVDF with GF Prefilter, 96-well plate, 1.0 m, PVDF with GF Prefilter, 96-well plate, 1.0 ml, Omni Screen Filter Plates, 96-we ll plate 1 * 10 ST
Catalog # MSPPOMNPF02-GF
Frequently Bought Together