Skip to Header
Skip to Main Content
Skip to Footer
Advanced
Advanced
SDS
Certificates
Manufacturer Data
0
SFr. 0.00
Order Entry
Login
/
Register
Products
Services
Digital Solutions
Applications
Support
About
Order Entry
Order Status
Your Account
Cart
Switzerland
Order Entry
Order Status
Your Account
Cart
ContactUsLinkComponent
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 20.40 MPA.S (25.00C), RTM10 1 * 500 mL
Catalog # PARGRTM10
Supplier:
PARAGON SCIENTIFIC
Certificates
Expand image
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 20.40 MPA.S (25.00C), RTM10 1 * 500 mL
Catalog # PARGRTM10
Supplier:
PARAGON SCIENTIFIC
Certificates
Related Information
Documentation
Certificates
Search by Lot Number
Documentation
Frequently Bought Together
Buffer solutions in sachets, Certipur®, Supelco®
Supplier: Merck
VWR® AVS TITRINORM Buffer Solutions, 20 °C
Supplier: VWR Chemicals
Ammonium standard solution, 1000 mg/L NH₄ in water (from NH₄Cl) Certipur® standard for Ion Chromatography, Supelco®
Supplier: Merck
Potassium hydrogen phthalate, Certipur® Reag. Ph. Eur., Reag. USP volumetric standard, secondary reference material for alkalimetry, Supelco®
Supplier: Merck
Buffer solutions, Certipur®, colour coded, Supelco®
Supplier: Merck
Ethyl acetate ≥99%, TECHNICAL
Supplier: VWR Chemicals
VWR®, Conductivity standard solutions
Supplier: VWR Chemicals
Buffer solutions, ready to use, referenced at 20 °C, Certipur®, Supelco®
Supplier: Merck
Potassium chloride 3 mol/l (3 N) in aqueous solution, Supelco®
Supplier: Merck
pH buffers, ready to use
Supplier: WTW
Acetonitrile, anhydrous (max. 0.003% H₂O) ≥99.95%, HiPerSolv CHROMANORM® Reag. USP, Reag. Ph. Eur., ACS, super gradient grade suitable for UPLC/UHPLC instruments
Supplier: VWR Chemicals
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 101.4 MPA.S (25.00C), RTM14 1 * 500 mL
Supplier: PARAGON SCIENTIFIC