ContactUsLinkComponent
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 2.144 MPA.S (25.00C), RTM4 1 * 500 mL
Catalog # PARGRTM4
Supplier:  PARAGON SCIENTIFIC
VISCOSITY STANDARD, ROTATIONAL MINERAL OIL, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 2.144 MPA.S (25.00C), RTM4 1 * 500 mL
Catalog # PARGRTM4
Supplier:  PARAGON SCIENTIFIC
Frequently Bought Together