ContactUsLinkComponent
VISCOSITY STANDARD, ROTATIONAL SILICONE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 100.7 MPA.S (25.00C), RT100 1 * 600 mL
Catalog # PARGVIS-RT100-600
Supplier:  PARAGON SCIENTIFIC
VISCOSITY STANDARD, ROTATIONAL SILICONE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 100.7 MPA.S (25.00C), RT100 1 * 600 mL
Catalog # PARGVIS-RT100-600
Supplier:  PARAGON SCIENTIFIC