VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 100.7 MPA.S (25 C) 1 * 500 mL
PARGS60
VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 100.7 MPA.S (25 C) 1 * 500 mL
PARGS60
Frequently Bought Together
VWR®, Specimen Containers, Conical, with Screw Caps
: VWR Collection
Acetone ≥99%, TECHNICAL
: VWR Chemicals
VWR® AVS TITRINORM Buffer Solutions, 20 °C
: VWR Chemicals
Single use syringes, 2-piece, Injekt® Solo
: B. Braun
VWR®, Nitrile Gloves
: VWR Collection
Sodium hydroxide standardised solution 0.05 N in aqueous solution
: Thermo Fisher Scientific
Yttrium, oil based standard solution, Y 1000 µg/g, Specpure®
: Thermo Fisher Scientific
Sodium Thiosulfate 0.1 mol/l (0.1N) (Reag. Ph.Eur.) volumetric solution 1 * 1 L
: PanReac AppliChem