VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 40049 MPA.S (25 C) 1 * 500 mL
Catalog # PARGN15000
Anbieter: PARAGON SCIENTIFIC
VISCOSITY STANDARD, GENERAL PURPOSE, UKAS ISO 17025 AND ISO 17034 CERTIFIED, NOMINAL VISCOSITY 40049 MPA.S (25 C) 1 * 500 mL
Catalog # PARGN15000
Anbieter: PARAGON SCIENTIFIC
Frequently Bought Together
Conductivity standards, KCl, referenced at 25 °C
Anbieter: REAGECON
VWR®, Silicone Rotational viscosity standards
Anbieter: VWR Chemicals
Supelco® Organic Reference Standard, Genistin
Anbieter: Merck
Supelco® Organic Reference Standard, Hexanoyl-ʟ-carnitine
Anbieter: Merck
Multi-element calibration standard 5
Anbieter: PerkinElmer
Colchicin 1 * 100 mg
Anbieter: EHRENSTORFER
CYCLIZINE-13C6 HCL 1 * 1 mL
Anbieter: Merck
Tetraconazole certified 1 * 0,1 g
Anbieter: EHRENSTORFER
UMAN STANDARD NF-L, LYOPHILIZED(X µl
Anbieter: MARKET SOURCE PART PROCESS
Petrolatum, white
Anbieter: USP
Polyethylene glycol 12,000, Supelco®
Anbieter: Merck