ContactUsLinkComponent
Liquid divider, with two glass needle valves, NS 29/32 inlet E, NS 14/23 oultlet A Spindle valves as locking valves (size SPV 6) and as ventilating valves (SPV 3) are used in the vacuum frame 1 * 1 items
Catalog # QVFLLSG0801031
Supplier:  QVF LABORTECHNIK
Liquid divider, with two glass needle valves, NS 29/32 inlet E, NS 14/23 oultlet A Spindle valves as locking valves (size SPV 6) and as ventilating valves (SPV 3) are used in the vacuum frame 1 * 1 items
Catalog # QVFLLSG0801031
Supplier:  QVF LABORTECHNIK
Frequently Bought Together