ContactUsLinkComponent
EL filling funnel for 10 lt canister - hard polyethylene (PE-HD). Made of black, electrically conductive or conductive PE-HD, with self-closing ball float, overfill protection and splash guard. Installation height 325mm, Ø 200mm, for canister Art. 10 1 * 1 items
Catalog # SEMA10568
Supplier:  SEMADENI
EL filling funnel for 10 lt canister - hard polyethylene (PE-HD). Made of black, electrically conductive or conductive PE-HD, with self-closing ball float, overfill protection and splash guard. Installation height 325mm, Ø 200mm, for canister Art. 10 1 * 1 items
Catalog # SEMA10568
Supplier:  SEMADENI
Frequently Bought Together