C1 Wiper Polyester Cellulose cleanroom wipe ultra-low tendency to generate particles and its ultra-low levels of soluble extractables. made of a super absorbent blend of cellulose and polyester. It is used in sheets for general purpose cleaning of eq 1 * 300 items
Catalog # EPTA7-C1-99P-00
Supplier: ESSENTRA POROUS TECHNOLOGIES
Frequently Bought Together