ContactUsLinkComponent
for HWS sliding ring stirrer cap Ceramic / high performance graphite, for 10 mm shaft-Ø Perfluoric gasket (Kalrez or similar) ***Attention: - no liability in case of improper installation*** 1 * 1 items
Catalog # HWSA3792010K
Supplier:  HWS LABORTECHNIK
for HWS sliding ring stirrer cap Ceramic / high performance graphite, for 10 mm shaft-Ø Perfluoric gasket (Kalrez or similar) ***Attention: - no liability in case of improper installation*** 1 * 1 items
Catalog # HWSA3792010K
Supplier:  HWS LABORTECHNIK
Frequently Bought Together