ContactUsLinkComponent
Target Protein,,MB Clonality,,Polyclonal Storage/Stability,,-20°C/1 year Application,,WB,IHC,IF/ICC,ELISA Recommended Dilution,,WB,,1:500-1:2000 IHC,,1:50-1:200, IF/ICC 1:100-1:500 Concentration,,1 mg/ml Reactive Species,,Human,Mouse,Rat Host Species 1 * 200 µG
Catalog # GENOITA7419200U488
Supplier:  G-Biosciences
Target Protein,,MB Clonality,,Polyclonal Storage/Stability,,-20°C/1 year Application,,WB,IHC,IF/ICC,ELISA Recommended Dilution,,WB,,1:500-1:2000 IHC,,1:50-1:200, IF/ICC 1:100-1:500 Concentration,,1 mg/ml Reactive Species,,Human,Mouse,Rat Host Species 1 * 200 µG
Catalog # GENOITA7419200U488
Supplier:  G-Biosciences
Frequently Bought Together